missing translation for 'onlineSavingsMsg'
Learn More

Activin C/Inhibin beta C Antibody (MM0076-4N17), DyLight 594, Novus Biologicals™

Product Code. 18414675 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414675 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414675 Supplier Novus Biologicals Supplier No. NBP212011DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Activin C/Inhibin beta C Monoclonal antibody specifically detects Activin C/Inhibin beta C in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Activin C/Inhibin beta C
Applications Western Blot
Classification Monoclonal
Clone MM0076-4N17
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias inhibin beta C chain, IHBC, inhibin, beta C
Host Species Mouse
Immunogen Human recombinant Activin C/Inhibin beta C
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 3626
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.