missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activin C/Inhibin beta C Antibody (MM0076-4N17), Allophycocyanin, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Activin C/Inhibin beta C Monoclonal antibody specifically detects Activin C/Inhibin beta C in Human samples. It is validated for Western Blot
Specifications
Specifications
| Applications | Western Blot |
| Classification | Monoclonal |
| Gene Alias | inhibin beta C chain, IHBC, inhibin, beta C |
| Host Species | Mouse |
| Immunogen | Human recombinant Activin C/Inhibin beta C |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?