missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activin C/Inhibin beta C Antibody (MM0076-4N17), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12011AF488
This item is not returnable.
View return policy
Description
Activin C/Inhibin beta C Monoclonal specifically detects Activin C/Inhibin beta C in Human samples. It is validated for Western Blot.
Specifications
| Activin C/Inhibin beta C | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| INHBC | |
| Human recombinant Activin C/Inhibin beta C | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| MM0076-4N17 | |
| Western Blot | |
| inhibin beta C chain, IHBC, inhibin, beta C | |
| Mouse | |
| Protein G purified | |
| Cytokine Research | |
| 3626 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction