missing translation for 'onlineSavingsMsg'
Learn More

Activin C/Inhibin beta C Antibody (CL6916), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-76514

 View more versions of this product

Product Code. 18216809

  • £469.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Activin C/Inhibin beta C Monoclonal specifically detects Activin C/Inhibin beta C in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Activin C/Inhibin beta C
Monoclonal
Unconjugated
PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide
INHBC
This antibody was developed against Recombinant Protein corresponding to amino acids: TPRAGGQCPACGGPTLELESQRELLLDLAKRSILDKLHLTQRPTLNRPVSRAALRTALQHLHGVPQGALLEDNREQECEI
100 μL
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2b
Western Blot
CL6916
Western Blot 1 ug/ml
inhibin beta C chain, IHBC, inhibin, beta C
Mouse
Protein A purified
Cytokine Research
3626.0
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.