missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activin B/Inhibin beta B Antibody (MM0075-9M29), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12010AF532
This item is not returnable.
View return policy
Description
Activin B/Inhibin beta B Monoclonal specifically detects Activin B/Inhibin beta B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Activin B/Inhibin beta B | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| INHBB | |
| Human recombinant Activin B/Inhibin beta B | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0075-9M29 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| activin AB beta polypeptide, Activin beta-B chain, INHBB, inhibin beta B chain, inhibin, beta B, inhibin, beta B (activin AB beta polypeptide), Inhibin, beta-2 | |
| Mouse | |
| 0.1 mL | |
| 3625 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction