missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activin B/Inhibin beta B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92836-0.1ml
This item is not returnable.
View return policy
Description
Activin B/Inhibin beta B Polyclonal antibody specifically detects Activin B/Inhibin beta B in Human samples. It is validated for Western Blot
Specifications
| Activin B/Inhibin beta B | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| activin AB beta polypeptide, Activin beta-B chain, INHBB, inhibin beta B chain, inhibin, beta B, inhibin, beta B (activin AB beta polypeptide), Inhibin, beta-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 293-407 of human INHBB (NP_002184.2). GLECDGRTNLCCRQQFFIDFRLIGWNDWIIAPTGYYGNYCEGSCPAYLAGVPGSASSFHTAVVNQYRMRGLNPGTVNSCCIPTKLSTMSMLYFDDEYNIVKRDVPNMIVEECGCA | |
| 0.1 mL | |
| Cytokine Research, Stem Cell Signaling Pathway | |
| 3625 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction