missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activin A Rabbit anti-Mouse, Clone: 105, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90669-100UL
This item is not returnable.
View return policy
Description
Activin A Monoclonal specifically detects Activin A in Mouse samples. It is validated for ELISA.
Specifications
| Activin A | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 105 | |
| ELISA 1:5000-1:10000 | |
| activin AB alpha polypeptide, Activin beta-A chain, erythroid differentiation factor, Erythroid differentiation protein, follicle-stimulating hormone-releasing protein, FSH-releasing protein, inhibin beta A chain, inhibin beta A subunit, Inhibin, beta-1 | |
| Recombinant Mouse Activin A protein | |
| 100 μg | |
| Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Stem Cell Signaling Pathway | |
| 3624 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction