missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Activin A Monoclonal Antibody (KAA378_14E10), Invitrogen™

Mouse Monoclonal Antibody

£416.00 - £734.00

Specifications

Antigen Activin A
Clone KAA378_14E10
Concentration 1 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30210910
View Documents
Invitrogen™
MA555850
50 μL
£416.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30193719
View Documents
Invitrogen™
MA556230
200 μL
£734.00
200µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Activin A Monoclonal antibody specifically detects Activin A in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

Activin A
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
activin A; activin beta-A chain; BMP7; BMP-7; Bone morphogenetic protein 7; EDF; Eptotermin alfa; erythroid differentiation factor; erythroid differentiation protein; follicle-stimulating hormone-releasing protein; FRP; FSH-releasing protein; INHB; Inhba; inhibin B subunit; inhibin beta A chain; inhibin beta A protein; inhibin beta A subunit; inhibin beta-A; inhibin beta-A-subunit; inhibin beta-subunit; inhibin subunit beta A; inhibin, beta A; inhibin, beta A (activin A, activin AB alpha polypeptide); Inhibin, beta-1; OP1; OP-1; Osteogenic protein 1; unnamed protein product
Recombinant human INHBA (immunogen range is 311-426).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Inhba
KAA378_14E10
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human
P08476
3624
IgA
Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.