missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Activin A Monoclonal antibody specifically detects Activin A in Mouse samples. It is validated for ELISA
Specifications
Specifications
| Purification Method | Protein A purified |
| Research Discipline | Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Stem Cell Signaling Pathway |
| Primary or Secondary | Primary |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?