missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Activin A Monoclonal antibody specifically detects Activin A in Mouse samples. It is validated for ELISA
Specifications
Specifications
| Conjugate | Janelia Fluor 549 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Activin A (Accession#: NP_032406.1; Met1-Ser424). |
| Purification Method | Protein A purified |
| Gene ID (Entrez) | 3624 |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?