missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Activin A Monoclonal specifically detects Activin A in Mouse samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | Activin A |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 105 |
| Conjugate | HRP |
| Formulation | PBS |
| Gene Alias | activin AB alpha polypeptide, Activin beta-A chain, erythroid differentiation factor, Erythroid differentiation protein, follicle-stimulating hormone-releasing protein, FSH-releasing protein, inhibin beta A chain, inhibin beta A subunit, Inhibin, beta-1 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Activin A (NP_032406.1; Met1-Ser424). |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?