Learn More
Erlab Activated Carbon Filtration - Solvents - Solis 1800
Product Code. 18838904 Shop All Erlab Products
Change view
Click to view available options
Product Type:
Filter
Unit Size:
Each
Product Code. Product Type Unit Size
18838904 Filter Each
1 options
Product Code. 18838904 Supplier Erlab Supplier No. NORT000352/AS
  • ONLINE EXCLUSIVE PRICE
    £2095.00
    £1918.17 / Pack of 1
    Save £176.83
    8% Off
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Filter for your Biological Safety Cabinet.
  • Activated carbon filtration for your BSC
  • Type of chemicals: Solvents
  • Compatible model: BSC Solis 1800
Specifications
Product Type Filter