missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Activated Carbon Filter

Product Code. 17675945 Shop All asecos Products
Change view
Click to view available options
For Use With (Equipment):
UFA.025.030.XL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17675945 UFA.025.030.XL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17675945 Supplier asecos™ Supplier No. 40202
Only null left
Add to basket
Edge
Add to basket

Square activated carbon filter for use with Recirculating Air Filter System UFA.025.030.XL.

  • Filter
  • For model(s): UFA.025.030(.XL)
  • Activated carbon filter (angular)

Specifications

Dimensions (D x W x H) 282 x 298 x 101 mm
For Use With (Application) Retention of Solvent Vapors (Hydrocarbons)
For Use With (Equipment) UFA.025.030.XL
Includes 1 Filter, Manual and Filter Code
Weight (English) 11.95 lb.
Weight (Metric) 5.42 kg
Product Type Activated Carbon Filter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.