Learn More
ERLAB F Activated Carbon Filter
Click to view available options
Hood Type:
F Extraction
Unit Size:
Each
Description
- The Activated Carbon - F - Extraction filter specializes in adsorbing formaldehyde and aldehyde vapors
- Essential in pathology, histology, and biosafety labs using fixatives or disinfectants containing formaldehyde
- Offers high retention capacity and prevents exposure to toxic fumes
- Reduces odor and protects user health in enclosed environments
- Compatible with ductless filtration systems and easy to maintain
- Helps labs comply with occupational exposure limits for formaldehyde
Specifications
Specifications
| Product Type | Carbon Filter |
| Hood Type | F Extraction |