missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 97-013 Medium Duty Hi-Viz™ Gloves with Intercept™ Technology

Product Code. 16885260 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
11
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 16885260

Brand: Ansell 97013090

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ergonomically designed gloves reduce hand fatigue, while highly visible coloring on the back of the hand and palm side increases safety in low light conditions.

  • Highly reflective back keeps workers safe in low-visibility and dark environments
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • 13-gauge liner offers increases comfort for all day wear
  • Breathable fabric minimizes perspiration compared to leather
  • Ergonomic design provides naturally curved, relaxed fit to reduce hand fatigue
  • Enhances grip capabilities in wet and dry conditions
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Applications Industrial
Coating Material Polyurethane
Color Hi-vis Orange
Cut Resistance Rating ANSI A2
Hand Specific
Material Intercept Technology Yarn
Product Type Multi-activity Gloves
Size 9
EN388 Standard 4X32B
Glove Cuff Style Knit
Series 97-013
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.