missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 97-012 General Purpose Hi-Viz™ Gloves

Product Code. p-7215022 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
11
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 16815260

Brand: Ansell 97012070

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Gloves are engineered to combine all-day comfort, dexterity, enhanced grip, and protection. Specially designed to keep workers safe in low-light or low-visibility environments.

  • High visibility on back of the hand and palm side increases safety
  • 15-gauge liner minimizes perspiration and offers comfortable all day wear
  • Ergonomically designed forms provide naturally curved, relaxed fit to reduce hand fatigue
  • Provides dexterity for the ability to pick up small items
  • Enhances grip in wet and dry conditions
  • Silicone free
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Applications Industrial
Coating Material Nitrile
Color Hi-vis Orange
Cut Resistance Rating ANSI A1
Hand Specific
Latex Free Yes
Material Nylon
Product Type Multi-activity Gloves
Size 7
EN388 Standard 3131A
Series 97-012
Yarn Gauge 15
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.