missing translation for 'onlineSavingsMsg'
Learn More

Cytiva ActiPro Liquid Medium

Product Code. 15835261
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 15835261

Brand: Cytiva SH31039.01

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

HyClone ActiPro cell culture medium has been formulated to provide high yields of recombinant proteins in processes using Chinese hamster ovary (CHO) cell lines. The medium is chemically defined, animal-derived component-free (ADCF), and optimized for high-yield protein production in batch or fed-batch processes.

  • ActiPro medium does not contain any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol, ensuring batch-to-batch consistency and increased cell culture process efficiency.
  • ActiPro medium is intended for use in combination with HyClone ActiSM medium and Cell Boost 7a and Cell Boost 7b supplements as part of the ActiPro system of media and supplements designed with adaptation and production in mind.
  • The ActiPro system is suitable for general biomanufacturing with CHO cell lines such as CHO-GS, CHO-K1, CHO-DG44, and CHO-S.
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Quantity 500 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.