missing translation for 'onlineSavingsMsg'
Learn More

Actinomycin D (Red Crystals), Fisher BioReagents

Product Code. 10285203
Change view
Click to view available options
Quantity:
5 mg
10 mg
Packaging:
Poly Micro Tube
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10285203 10 mg Poly Micro Tube 10mg
10688203 5 mg Poly Micro Tube 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10285203 Supplier Fisher BioReagents Supplier No. BP60610
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Isolated from Streptomyces parvuius

This antibiotic is a widely used transcription inhibitor.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-76-0
Molecular Formula C62H86N12O16
Molecular Weight (g/mol) 1255.438
InChI Key RJURFGZVJUQBHK-HUZQGMAJSA-N
PubChem CID 131954673
IUPAC Name 2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,
SMILES CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C

Specifications

Type Actinomycin D
Physical Form Powder/Solid
Melting Point 245°C
Extinction Coefficient Report
Packaging Poly Micro Tube
Color Red
Quantity 10 mg
Assay Percent Range ≥98 %
Merck Index 15, 2797
Solubility Information Soluble in Methanol
Formula Weight 1255.44
Percent Purity 99.0% min. (HPLC), 98% min. (TLC)
Chemical Name or Material Actinomycin-D
Show More Show Less
compliance-icons
Product Identifier
  • Actinomycin D
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
Hazard Statement
  • H300-Fatal if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.