missing translation for 'onlineSavingsMsg'
Learn More

Actinomycin D (Dactinomycin), TRC

Product Code. 18010930
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18010930 10 mg 10mg
18081230 50 mg 50mg
18042330 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18010930 Supplier Toronto Research Chemicals Supplier No. TRCD10250010MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-76-0
Molecular Formula C62 H86 N12 O16
Molecular Weight (g/mol) 1255.42
Synonym Actinomycin D, 3H-Phenoxazine-1,9-dicarboxamide, 2-amino-N,N'-bis(hexadecahydro-6,13-diisopropyl-2,5,9-trimethyl-1,4,7,11,14-pentaoxo-1H-pyrrolo[2,1-i][1,4,7,10,13]oxatetraazacyclohexadecin-10-yl)-4,6-dimethyl-3-oxo- (7CI), Actinomycin C1 (6CI), 1H-Pyrrolo[2,1-i][1,4,7,10,13]oxatetraazacyclohexadecine, cyclic peptide deriv., 3H-Phenoxazine, actinomycin D deriv., AD, Actactinomycin A IV, Actinomycin 7, Actinomycin AIV, Actinomycin I1, Actinomycin IV, Actinomycin X1, Actinomycindioic D acid, dilactone, C 1, Chounghwamycin B, Cosmegen, Dactinomycin, Dactinomycin D, Dilactone actinomycin D acid, Dilactone actinomycindioic D acid, Lyovac cosmegen, Meractinomycin, NCI C04682, NSC 3053, Oncostatin K
IUPAC Name 2-amino-4,6-dimethyl-3-oxo-1-N,9-N-bis[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]phenoxazine-1,9-dicarboxamide
SMILES CC(C)[C@H]1NC(=O)[C@@H](NC(=O)C2=C(N)C(=O)C(=C3Oc4c(C)ccc(C(=O)N[C@H]5[C@@H](C)OC(=O)[C@H](C(C)C)N(C)C(=O)CN(C)C(=O)[C@@H]6CCCN6C(=O)[C@H](NC5=O)C(C)C)c4N=C23)C)[C@@H](C)OC(=O)[C@H](C(C)C)N(C)C(=O)CN(C)C(=O)[C@@H]7CCCN7C1=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C62H86N12O16/c1-27(2)42-59(84)73-23-17-19-36(73)57(82)69(13)25-38(75)71(15)48(29(5)6)61(86)88-33(11)44(55(80)65-42)67-53(78)35-22-21-31(9)51-46(35)64-47-40(41(63)50(77)32(10)52(47)90-51)54(79)68-45-34(12)89-62(87)49(30(7)8)72(16)39(76)26-70(14)58(83)37-20-18-24-74(37)60(85)43(28(3)4)66-56(45)81/h21-22,27-30,33-34,36-37,42-45,48-49H,17-20,23-26,63H2,1-16H3,(H,65,80)(H,66,81)(H,67,78)(H,68,79)/t33-,34-,36+,37+,42-,43-,44+,45+,48+,49+/m1/s1
Formula Weight 1254.6285
Percent Purity >95%
Chemical Name or Material Dactinomycin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.