missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Actinomycin D

Product Code. 11524137
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11524137

Brand: Invitrogen™ A7592

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Actinomycin D exhibits high GC selectivity and causes distortion at its binding site. Binding of the nonfluorescent actinomycin D to nucleic acids changes the absorbance of the dye. Actinomycin D has been used for chromosome banding studies.

Cell Analysis, Cell Proliferation, Cell Viability, Proliferation & Function

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Violet
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Colorimetric
Form Solid
Product Type Antibiotic
Quantity 10 mg
Shipping Condition Room Temperature
compliance-icons compliance-icons
Product Identifier
  • actinomycin D
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Carcinogenicity Category 1
  • Reproductive toxicity Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Actinomycin D

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.