missing translation for 'onlineSavingsMsg'
Learn More

Melford Actinomycin D

Product Code. 30017927
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30017927 10 mg 10mg
30017926 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30017927 Supplier Melford Supplier No. A100250.010
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Actinomycin D, also known as Dactinomycin, is an antibiotic.

Actinomycin D, also known as Dactinomycin, is an antibiotic.

It works by binding to DNA, inhibiting both DNA and RNA synthesis. This interference with nucleic acid synthesis makes it effective against rapidly dividing cells, such as cancer cells.

Actinomycin D is used as a tool to study DNA and RNA synthesis, cell cycle progression, and cellular responses to DNA damage. Its ability to interfere with nucleic acid synthesis makes it valuable in understanding the molecular mechanisms of cell biology and cancer.

Specifications

Chemical Name or Material Actinomycin D [Actinomycin C1] [Dactinomycin]
CAS 50-76-0
Quantity 10 mg
Molecular Formula C62H86N12O16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.