missing translation for 'onlineSavingsMsg'
Learn More
Learn More
actin-related protein 2/3 complex subunit 1B Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99248AF647
This item is not returnable.
View return policy
Description
actin-related protein 2/3 complex subunit 1B Polyclonal antibody specifically detects actin-related protein 2/3 complex subunit 1B in Human samples. It is validated for Western Blot
Specifications
| actin-related protein 2/3 complex subunit 1B | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 10095 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 647 | |
| actin related protein 2/3 complex, subunit 1B (41 kD), actin related protein 2/3 complex, subunit 1B, 41kDa, actin-related protein 2/3 complex subunit 1B, ARC41p41-ARCp40-ARC, Arp2/3 complex 41 kDa subunit, ARP2/3 protein complex subunit p41 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human actin-related protein 2/3 complex subunit 1B. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction