missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Actin (Rabbit)

Product Code. 16161750
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 16161750

Brand: Abnova™ P5204.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for SDS-PAGE

#REF!

Specifications

Accession Number P68135.1
For Use With (Application) SDS-PAGE
Molecular Weight (g/mol) 42kDa
Name Actin (Rabbit)
Preparation Method Native protein purified from Rabbit Muscle by modification of the procedure of Spudich and Watt, J. Biol. Chem., 246, 4866 (1971)
Quantity 1 mg
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name ACTIN
Assays Binding studies; Endogenous internal controls to normalize gene expression studies.
Species Rabbit
Protein Tag None
Expression System Native protein purified from Rabbit Muscle by modification of the procedure of Spudich and Watt, J. Biol. Chem., 246, 4866 (1971)
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.