missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actin (Muscle Specific) Mouse anti-Human, Rat, Rabbit, Clone: rMSA/953, Novus Biologicals™
Mouse Monoclonal Antibody
£197.00 - £396.00
Specifications
| Antigen | Actin (Muscle Specific) |
|---|---|
| Clone | rMSA/953 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 0.5-1.0 ug/ml, Immunohistochemistry-Paraffin 0.5-2 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18352877
|
Novus Biologicals
NBP3-13908-20UG |
20 μg |
£197.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18331684
|
Novus Biologicals
NBP3-13908-100UG |
100 μg |
£396.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Actin (Muscle Specific) Monoclonal antibody specifically detects Actin (Muscle Specific) in Human, Rat, Rabbit samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
| Actin (Muscle Specific) | |
| 0.2 mg/ml | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Developmental Biology, Extracellular Matrix, Signal Transduction, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| ACTA, ACTC, actin, alpha 2, smooth muscle, aorta, actin, alpha cardiac muscle 1, actin, alpha skeletal muscle, actin, aortic smooth muscle, ACTSA, alpha-cardiac actin, ASD5, ASMA, Cell growth-inhibiting gene 46 protein, CFTD, CFTD1, CFTDM, CMD1R, CMH11, MPFD, NEM1, NEM2, NEM3, nemaline myopathy type 3 | |
| Recombinant protein corresponding to human muscle actin | |
| Primary | |
| Store at 4°C. |
| rMSA/953 | |
| Western Blot 0.5-1.0 ug/ml, Immunohistochemistry-Paraffin 0.5-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat, Rabbit | |
| P62736, P68133, P68032 | |
| 58 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title