missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actin (Muscle Specific) Antibody (ACTA1/9105R), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-24312CL1
This item is not returnable.
View return policy
Description
Actin (Muscle Specific) Monoclonal antibody specifically detects Actin (Muscle Specific) in Human samples. It is validated for Flow Cytometry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| Actin (Muscle Specific) | |
| Monoclonal | |
| CoraFluor 1 | |
| ACTA, ACTC, actin, alpha 2, smooth muscle, aorta, actin, alpha cardiac muscle 1, actin, alpha skeletal muscle, actin, aortic smooth muscle, ACTSA, alpha-cardiac actin, ASD5, ASMA, Cell growth-inhibiting gene 46 protein, CFTD, CFTD1, CFTDM, CMD1R, CMH11, MPFD, NEM1, NEM2, NEM3, nemaline myopathy type 3 | |
| Recombinant human Actin (Muscle Specific) protein | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Developmental Biology, Extracellular Matrix, Signal Transduction, Stem Cell Markers | |
| 58 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG κ |
| Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| ACTA1/9105R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction