missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTIN Mouse anti-Chicken, Human, Mouse, Rat, Clone: AC-40, Abnova™
Mouse monoclonal antibody raised against partial synthetic peptide of actin.
Brand: Abnova MAB12450.100ug
This item is not returnable.
View return policy
Description
Sequence: SGPSIVHRKCFSpecifications
| ACTIN | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (4 ug/mL) Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Store at −20°C. After reconstitution with 1mLof 1.2% sodium acetate or neutral PBS and concentration will be 100μg/mL, store at −20°C. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a |
| Immunohistochemistry (Frozen), Western Blot | |
| AC-40 | |
| Mouse monoclonal antibody raised against partial synthetic peptide of actin. | |
| Lyophilized from ascites fluid (1.2% sodium acetate, 2mg BSA, 0.01mg sodium azide) | |
| A synthetic peptide corresponding to at the C-terminus of actin. (Multiple Antigen Peptide backbone) | |
| RUO | |
| Chicken, Human, Mouse, Rat | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction