missing translation for 'onlineSavingsMsg'
Learn More

Actin Gamma 1 Antibody (2A3), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18407047 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407047 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407047 Supplier Novus Biologicals Supplier No. NBP197720MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Actin Gamma 1 Monoclonal antibody specifically detects Actin Gamma 1 in Human, Mouse, Rat, Porcine, Chicken, Rabbit, Zebrafish samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Actin Gamma 1
Applications Flow Cytometry
Classification Monoclonal
Clone 2A3
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias ACT, ACTB, ACTG, actin, cytoplasmic 2, actin, gamma 1, cytoskeletal gamma-actin, deafness, autosomal dominant 20; deafness, autosomal dominant 26, DFNA20, DFNA26, Gamma-actin
Host Species Mouse
Immunogen Derived by fusion of NS0 Mouse myeloma cells with spleen cells from a BALB/c Mouse immunized with a peptide comprising the N-terminal nonapeptide of gamma-cytoplasmic actin with an acetylated N-terminus (Ac-EEEIAALVI-COOH) coupled to keyhole limpet hemocyanin through the cysteine residue. The antibody does not cross react with other actin isoforms.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 71
Target Species Human, Mouse, Rat, Pig, Chicken, Rabbit, Zebrafish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.