missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTH Rabbit anti-Human, Clone: CLIP/2859R, Novus Biologicals™
Rabbit Monoclonal Antibody
£431.00
Specifications
| Antigen | ACTH |
|---|---|
| Clone | CLIP/2859R |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA |
| Applications | ELISA |
Description
ACTH Monoclonal specifically detects ACTH in Human samples. It is validated for ELISA.Specifications
| ACTH | |
| 0.2 mg/mL | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| 10 mM PBS with 0.05% BSA | |
| 5443 | |
| IgG | |
| Protein A or G purified |
| CLIP/2859R | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein | |
| Synthetic peptide corresponding to aa25-39 of human ACTH (NGAEDESAEAFPLEF) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title