missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTA2/smooth muscle actin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ACTA2/smooth muscle actin |
|---|---|
| Concentration | 0.29 mg/mL |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16853014
|
Proteintech
14395-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16843014
|
Proteintech
14395-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody may cross-react with other isoforms of actins.
Among the six actin isoforms described in mammals, two are found in virtually all cells (b- and g-cytoplasmic), two are detected in smooth muscle cells (a- and g-smooth muscle) and two are present in striated muscles, one predomitly in skeletal (a-skeletal) and one in cardiac (a-cardiac) muscle cells. These actin isoforms differ slightly in their N-terminus and the sequences of each of them are perfectly conserved in higher vertebrates. Alpha-smooth muscle actin is abundant in vascular and visceral smooth muscle cells. In addition, it has also been shown to appear in stress fibers of fibroblastic cells during pathological situations involving contractile phenomena such as wound healing and fibrocontractive diseases.Specifications
| ACTA2/smooth muscle actin | |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| P62736, P62737, P62738 | |
| 11475, 59, 81633 | |
| ACTA2/smooth muscle actin Fusion Protein Ag5886 | |
| Primary | |
| -20°C | |
| ACTA2 |
| 0.29 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| a SMA, AAT6, ACTA2, ACTA2 Smooth muscle Actin, Actin, aortic smooth muscle, Actin, aortic smooth muscle Actin, ACTSA, ACTVS, Alpha actin 2, Alpha SMA, a-SMA, alpha SMA | |
| ACTA2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title