missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACSS3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566398
This item is not returnable.
View return policy
Description
Immunogen sequence: MDEEGYLYVMS RVDDVINVAG HRISAGAIEE SILSHGTVAD CAVVGKEDPL KGHVPLALCV LRKDINATEE QVLEEIVKHV RQNIGPVAAF RNAVFVK Highest antigen sequence identity to the following orthologs - mouse 95%, rat 95%.
Catalyzes the synthesis of acetyl-CoA from short-chain fatty acids (PubMed:28003429). Propionate is the preferred substrate (PubMed:28003429). Can utilize acetate and butyrate with a much lower affinity. [UniProt].
Specifications
| ACSS3 | |
| Polyclonal | |
| Unconjugated | |
| Acss3 | |
| 8430416H19Rik; Acetate--CoA ligase 3; ACSS3; Acyl-CoA synthetase short-chain family member 3; acyl-CoA synthetase short-chain family member 3, mitochondrial; AMP-binding enzyme, 33217; Gm874; Propionate--CoA ligase; RGD1307051 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79611 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H6R3 | |
| Acss3 | |
| Recombinant Human ACSS3. Recombinant protein control fragment (Product #RP-105193). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction