missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACSS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559392
This item is not returnable.
View return policy
Description
Immunogen sequence: IFAGFSAESL AGRINDAKCK VVITFNQGLR GGRVVELKKI VDEAVKHCPT VQHVLVAHRT DNKVHMGDLD VPLEQEMAKE DPVCAPESMG SEDMLFML Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.
This gene encodes a mitochondrial acetyl-CoA synthetase enzyme. A similar protein in mice plays an important role in the tricarboxylic acid cycle by catalyzing the conversion of acetate to acetyl CoA. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
| ACSS1 | |
| Polyclonal | |
| Unconjugated | |
| ACSS1 | |
| 1110032O15Rik; Acas2; ACAS2L; ACECS1; AceCS2; AceCS2L; acetate--CoA ligase 2; Acetyl-CoA synthetase 2; acetyl-coenzyme A synthetase 2-like, mitochondrial; ACSS1; Acyl-CoA synthetase short-chain family member 1; AI788978; KIAA1846; Propionate--CoA ligase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84532 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NUB1 | |
| ACSS1 | |
| Recombinant protein corresponding to Human ACSS1. Recombinant protein control fragment (Product #RP-99451). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction