missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACSM3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559391
This item is not returnable.
View return policy
Description
Immunogen sequence: KANCIITNDV LAPAVDAVAS KCENLHSKLI VSENSREGWG NLKELMKHAS DSHTCVKTKH NEIMAIFF Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 81%.
Has medium-chain fatty acid:CoA ligase activity with broad substrate specificity (in vitro). Acts on acids from C(4) to C(11) and on the corresponding 3-hydroxy- and 2,3- or 3,4-unsaturated acids (in vitro).
Specifications
| ACSM3 | |
| Polyclonal | |
| Unconjugated | |
| ACSM3 | |
| Acsm3; acyl-CoA synthetase medium chain family member 3; acyl-CoA synthetase medium-chain family member 3; Acyl-coenzyme A synthetase ACSM3, mitochondrial; butyrate--CoA ligase 3; butyryl coenzyme A synthetase 3; butyryl-coenzyme A synthetase 3; CH73-60L17.3; Middle-chain acyl-CoA synthetase 3; Propionate--CoA ligase; Protein SA; protein SA homolog; Sa; SA (rat hypertension-associated) homolog; SA hypertension-associated homolog; SA rat hypertension-associated homolog; SA rat hypertension-associated protein; SAH; ssubA; zgc:172040 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6296 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q53FZ2 | |
| ACSM3 | |
| Recombinant protein corresponding to Human ACSM3. Recombinant protein control fragment (Product #RP-98670). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction