missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Target | ACSM3 |
| Synonym | acyl-CoA synthetase medium-chain family member 3Protein SA homolog, acyl-coenzyme A synthetase ACSM3, mitochondrial, Butyrate--CoA ligase 3, Butyryl-coenzyme A synthetase 3, EC 6.2.1, Middle-chain acyl-CoA synthetase 3, SA, SA (rat hypertension-associated) homolog, SA hypertension-associated homolog, SA hypertension-associated homolog (rat), SAH, SAHEC 6.2.1.2 |
| Gene Symbol | ACSM3 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?