missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACSM2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570859
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 85%; Dog: 93%; Guinea Pig: 86%; Horse: 85%; Human: 100%; Mouse: 86%; Rabbit: 93%; Rat: 90%; Zebrafish: 85%.
This gene encodes a mitochondrial acyl-coenzyme A synthetase that is specific for medium chain fatty acids. These enzymes catalyze fatty acid activation, the first step of fatty acid metabolism, through the transfer of acyl-CoA. These enzymes also participate in the glycine conjugation pathway in the detoxification of xenobiotics such as benzoate and ibuprofen. Expression levels of this gene in the kidney may be correlated with kidney function. This gene and its paralog ACSM2B (Gene ID: 348158), both present on chromosome 16, likely arose from a chromosomal duplication event.
Specifications
| ACSM2A | |
| Polyclonal | |
| Unconjugated | |
| ACSM2A | |
| A-923A4.1; ACSM2; ACSM2A; acyl-CoA synthetase medium-chain family member 2; acyl-CoA synthetase medium-chain family member 2A; Acyl-coenzyme A synthetase ACSM2A, mitochondrial; Benzoate--CoA ligase; Butyrate--CoA ligase 2A; butyryl-coenzyme A synthetase 2A; Homolog of rat kidney-specific (KS); MACS2; Middle-chain acyl-CoA synthetase 2A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 123876 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q08AH3 | |
| ACSM2A | |
| synthetic peptide directed towards the C-terminal region of human ACSM2A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction