missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACSM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145454
This item is not returnable.
View return policy
Description
ACSM1 has medium-chain fatty acid:CoA ligase activity with broad substrate specificity (in vitro). This protein acts on acids from C(4) to C(11) and on the corresponding 3-hydroxy-and 2,3-or 3,4-unsaturated acids (in vitro). This protein functions as GTP-dependent lipoate-activating enzyme that generates the substrate for lipoyltransferase.
Specifications
| ACSM1 | |
| Polyclonal | |
| Unconjugated | |
| ACSM1 | |
| ACSM1; Acyl-CoA synthetase medium-chain family member 1; acyl-coenzyme A synthetase ACSM1, mitochondrial; Benzoate--CoA ligase; BUCS1; Butyrate CoA ligase; butyrate--CoA ligase 1; butyryl Coenzyme A synthetase 1; butyryl-coenzyme A synthetase 1; LAE; Lipoate-activating enzyme; MACS1; medium-chain acyl-CoA synthetase; middle-chain acyl-CoA synthetase 1; Xenobiotic/medium-chain fatty acid-CoA ligase HXM-B | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 116285, 117147, 361638 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Q08AH1, Q91VA0 | |
| ACSM1 | |
| Recombinant Human ACSM1 protein, Ser370-Pro557 (Accession #Q08AH1). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction