missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSL5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15708-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. This isozyme is highly expressed in uterus and spleen, and in trace amounts in normal brain, but has markedly increased levels in maligt gliomas. This gene functions in mediating fatty acid-induced glioma cell growth. Three transcript variants encoding two different isoforms have been found for this gene.Specifications
| ACSL5 | |
| Polyclonal | |
| Unconjugated | |
| ACSL5 | |
| ACS2, ACS5, ACSL5, FACL5, LACS 5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 433256, 51703, 94340 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88813, Q8JZR0, Q9ULC5 | |
| ACSL5 | |
| ACSL5 Fusion Protein Ag8258 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction