missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSL5 Antibody (CL0275) [Alexa Fluor™ 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52899AF594
This item is not returnable.
View return policy
Description
ACSL5 Monoclonal antibody specifically detects ACSL5 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| ACSL5 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| ACS2, ACS5FACL5 for fatty acid coenzyme A ligase 5, acyl-CoA synthetase long-chain family member 5, EC 6.2.1, EC 6.2.1.3, FACL5fatty acid coenzyme A ligase 5, fatty-acid-Coenzyme A ligase, long-chain 5, LACS 5, Long-chain acyl-CoA synthetase 5, long-chain fatty acid coenzyme A ligase 5, long-chain-fatty-acid--CoA ligase 5 | |
| This antibody was developed using a recombinant protein derived from Q9ULC5, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism | |
| 51703 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL0275 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction