missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSL4/FACL4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22401-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes the second largest subunit of RNA polymerase III, the polymerase responsible for synthesizing transfer and small ribosomal RNAs in eukaryotes. The largest subunit and the encoded protein form the catalytic center of RNA polymerase III. Mutations in this gene are a cause of hypomyelinating leukodystrophy. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Specifications
| ACSL4/FACL4 | |
| Polyclonal | |
| Unconjugated | |
| ACSL4 | |
| ACS4, ACSL4, FACL4, LACS 4, LACS4, MRX63, MRX68 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2182 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60488 | |
| ACSL4 | |
| ACSL4/FACL4 Fusion Protein Ag18085 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction