missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ACSL1 Polyclonal antibody specifically detects ACSL1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | ACSL1 |
| Applications | Western Blot |
| Host Species | Rabbit |
| Immunogen | Rabbit polyclonal ACSL1 antibody was raised against an 18 amino acid peptide near the center of human ACSL1. The immunogen is located within amino acids 240 - 290 of ACSL1. Amino Acid Squence: CDLGRANRRKPKPPAPEDL |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Research Discipline | Cancer, Lipid and Metabolism, Signal Transduction |
| Primary or Secondary | Primary |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?