missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSF3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £358.00
Specifications
| Antigen | ACSF3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
ACSF3 Polyclonal antibody specifically detects ACSF3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| ACSF3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS with 50% glycerol, pH7.3. | |
| 197322 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| acyl-CoA synthetase family member 3, acyl-CoA synthetase family member 3, mitochondrial, EC 6.2.1, EC 6.2.1.-, FLJ39242 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 270-450 of human ACSF3 (NP_001230208.1). FSPQQVWEKFLSSETPRINVFMAVPTIYTKLMEYYDRHFTQPHAQDFLRAVCEEKIRLMVSGSAALPLPVLEKWKNITGHTLLERYGMTEIGMALSGPLTTAVRLPGSVGTPLPGVQVRIVSENPQREACSYTIHAEGDERGTKVTPGFEEKEGELLVRGPSVFREYWNKPEETKSAFTLD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title