missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACSBG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559659
This item is not returnable.
View return policy
Description
Immunogen sequence: ANVIMVDTQK QLEKILKIWK QLPHLKAVVI YKEPPPNKMA NVYTMEEFME LGNEVPEEAL DAIIDTQQPN QCCVLV Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
The protein encoded by this gene possesses long-chain acyl-CoA synthetase activity. It is thought to play a central role in brain very long-chain fatty acids metabolism and myelinogenesis.
Specifications
| ACSBG1 | |
| Polyclonal | |
| Unconjugated | |
| ACSBG1 | |
| ACSBG1; Acyl-CoA synthetase bubblegum family member 1; BG; BG1; BG1 protein; BGM; bubblegum; E230019G03Rik; Gonadotropin-regulated long chain acyl CoA synthetase; gonadotropin-regulated long chain acyl-CoA synthetase; Grlacs; GR-LACS; hBG1; KIAA0631; Lipidosin; lipidosis-related protein lipidosin; lipodisin; long-chain-fatty-acid--CoA ligase ACSBG1; LPD; mBG1; mKIAA0631; R75185; very long-chain acyl-CoA synthetase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23205 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96GR2 | |
| ACSBG1 | |
| Recombinant protein corresponding to Human ACSBG1. Recombinant protein control fragment (Product #RP-98019). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction