missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACSBG1 Monoclonal Antibody (OTI2B5)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525103
This item is not returnable.
View return policy
Description
ACSBG1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene possesses long-chain acyl-CoA synthetase activity. It is thought to play a central role in brain very long-chain fatty acids metabolism and myelinogenesis.
Specifications
| ACSBG1 | |
| Monoclonal | |
| 0.95 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96GR2 | |
| ACSBG1 | |
| Full length human recombinant protein of ACSBG1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI2B5 | |
| Unconjugated | |
| ACSBG1 | |
| ACSBG1; Acyl-CoA synthetase bubblegum family member 1; BG; BG1; BG1 protein; BGM; bubblegum; E230019G03Rik; Gonadotropin-regulated long chain acyl CoA synthetase; gonadotropin-regulated long chain acyl-CoA synthetase; Grlacs; GR-LACS; hBG1; KIAA0631; Lipidosin; lipidosis-related protein lipidosin; lipodisin; long-chain-fatty-acid--CoA ligase ACSBG1; LPD; mBG1; mKIAA0631; R75185; very long-chain acyl-CoA synthetase | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23205 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction