missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSBG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-58214
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
ACSBG1 Polyclonal specifically detects ACSBG1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Spécification
| ACSBG1 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| acyl-CoA synthetase bubblegum family member 1EC 6.2.1.3, BG1, BGMBG, bubblegum, FLJ30320, hBG1, hsBG, hsBGM, KIAA0631GR-LACS, lipidosin, long-chain-fatty-acid--CoA ligase ACSBG1, LPD, MGC14352, very long-chain acyl-CoA synthetase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| ACSBG1 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TLKCTLDPDTSDQTDNLTEQAVEFCQRVGSRATTVSEIIEKKDEAVYQAIEEGIRRVNMNAAARPYHIQKWAILER | |
| 100 μL | |
| Lipid and Metabolism | |
| 23205 | |
| Human | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu