missing translation for 'onlineSavingsMsg'
Learn More

Acryloyl Chloride, TRC

Product Code. 30006941
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006941 100 g 100g
30009818 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006941 Supplier Toronto Research Chemicals Supplier No. TRCA191395100G
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 814-68-6
Molecular Formula C3H3ClO
Molecular Weight (g/mol) 90.51
Synonym 2-Propenoyl Chloride, Acrylic Acid Chloride, Acrylyl Chloride, NSC 93770, Propenoyl Chloride
IUPAC Name prop-2-enoyl chloride
SMILES ClC(=O)C=C

Specifications

Quantity 100 g
InChI Formula InChI=1S/C3H3ClO/c1-2-3(4)5/h2H,1H2
Formula Weight 89.9872
Chemical Name or Material Acryloyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.