missing translation for 'onlineSavingsMsg'
Learn More

Acryloyl chloride, 96%, stab. with 400ppm phenothiazine

Product Code. p-7045347
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11308395

Brand: Thermo Scientific Alfa Aesar L10363.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 814-68-6 | C3H3ClO | 90.51 g/mol

Acryloyl chloride is used in the production of plastics. It plays an important role in the preparation of acrylate monomers and polymers. It also acts as a substrate for cross-metathesis. Further, it is utilized in organic synthesis for the introduction of acrylic groups into other compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acryloyl chloride is used in the production of plastics. It plays an important role in the preparation of acrylate monomers and polymers. It also acts as a substrate for cross-metathesis. Further, it is utilized in organic synthesis for the introduction of acrylic groups into other compounds.

Solubility
Miscible with water.

Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with alcohols, oxidizing agents, strong bases and polymerizing initiators.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 814-68-6
Molecular Formula C3H3ClO
Molecular Weight (g/mol) 90.51
MDL Number MFCD00000731
InChI Key HFBMWMNUJJDEQZ-UHFFFAOYSA-N
Synonym acryloyl chloride, acrylyl chloride, 2-propenoyl chloride, acrylic acid chloride, propenoyl chloride, acryloylchloride, chlorid kyseliny akrylove, unii-8k23o56tg5, chlorid kyseliny akrylove czech, acrylylchloride
PubChem CID 13140
IUPAC Name prop-2-enoyl chloride
SMILES ClC(=O)C=C

Specifications

Density 1.114
Boiling Point 74°C to 76°C
Flash Point −4°C (24°F)
Odor Acrid
Linear Formula CH2=CHCOCl
Refractive Index 1.435
Quantity 10 g
UN Number 3383
Beilstein 635744
Sensitivity Light and moisture sensitive
Solubility Information Miscible with water.
Formula Weight 90.51
Percent Purity 96%
Chemical Name or Material Acryloyl Chloride, stabilized with 400ppm phenothiazine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Acryloyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • EUH014-Reacts violently with water.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Acrylic acid chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.