missing translation for 'onlineSavingsMsg'
Learn More

Acryloyl chloride, 96%, stab. with 400ppm phenothiazine

Product Code. 11328395
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11328395 250 g 250g
11308395 10 g 10g
11318395 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11328395 Supplier Thermo Scientific Alfa Aesar Supplier No. L10363.30
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 814-68-6 | C3H3ClO | 90.51 g/mol

Acryloyl chloride is used in the production of plastics. It plays an important role in the preparation of acrylate monomers and polymers. It also acts as a substrate for cross-metathesis. Further, it is utilized in organic synthesis for the introduction of acrylic groups into other compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acryloyl chloride is used in the production of plastics. It plays an important role in the preparation of acrylate monomers and polymers. It also acts as a substrate for cross-metathesis. Further, it is utilized in organic synthesis for the introduction of acrylic groups into other compounds.

Solubility
Miscible with water.

Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with alcohols, oxidizing agents, strong bases and polymerizing initiators.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 814-68-6
Molecular Formula C3H3ClO
Molecular Weight (g/mol) 90.51
MDL Number MFCD00000731
InChI Key HFBMWMNUJJDEQZ-UHFFFAOYSA-N
Synonym acryloyl chloride, acrylyl chloride, 2-propenoyl chloride, acrylic acid chloride, propenoyl chloride, acryloylchloride, chlorid kyseliny akrylove, unii-8k23o56tg5, chlorid kyseliny akrylove czech, acrylylchloride
PubChem CID 13140
IUPAC Name prop-2-enoyl chloride
SMILES ClC(=O)C=C

Specifications

Density 1.114
Boiling Point 74°C to 76°C
Flash Point −4°C (24°F)
Odor Acrid
Linear Formula CH2=CHCOCl
Refractive Index 1.435
Quantity 250 g
UN Number UN3383
Beilstein 635744
Sensitivity Light and moisture sensitive
Solubility Information Miscible with water.
Formula Weight 90.51
Percent Purity 96%
Chemical Name or Material Acryloyl chloride, stabilized with 400ppm phenothiazine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Acryloyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • EUH014-Reacts violently with water.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Acryloyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.