missing translation for 'onlineSavingsMsg'
Learn More

Deutsch Neumann™ Acrylnitril-Butadien Stoppers

Product Code. 10411104
Change view
Click to view available options
Diameter (Metric) Bottom:
18 mm
21 mm
23 mm
29 mm
31 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Bottom unitSize
10411104 23 mm Each
10340671 21 mm Each
10579412 29 mm Each
10014770 18 mm Each
10606371 31 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10411104 Supplier Deutsch Neumann™ Supplier No. 1010323
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Hardness 45 Shore A
Height (Metric) 30mm
Quantity 1/Pk.
Color Black
Diameter (Metric) Bottom 23 mm
Diameter (Metric) Top 29 mm
Material Acrylonitrile Butadiene
Product Type Tapered Stopper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.