missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sanplatec™ Acrylic Vacuum Desiccator YB-1 Type
Description
- All-acrylic vacuum desiccator with excellent visibility and airtightness.
- The shelves are removable, and the number of shelves can be increased or decreased depending on the purpose.
- Vacuum gauge 1pcs for 0 - 0.1Mpa , Valve 2pcs, nozzle O.D. 9 mm, Gasket: Silicone
Specifications
Specifications
| Material | Acrylic |
| Product Type | Vacuum Desiccator |
| Volume (Metric) | 14 L |
| Height (Metric) | 380 mm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?