missing translation for 'onlineSavingsMsg'
Learn More

Sanplatec™ Acrylic Vacuum Desiccator Portable SB-P Type

Product Code. 30245639
Change view
Click to view available options
Height (Metric):
386mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) unitSize
30245639 386mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30245639 Supplier Sanplatec™ Supplier No. 230+BS/RCSB2P
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Transparent acrylic allows great visibility with top opening. Compact type.

  • Vacuum gauge 1pc for 0 - 0.1Mpa
  • Valve 2pcs, nozzle O.D. 9 mm
  • Gasket: Silicone

Specifications

Material Acrylic
Product Type Vacuum Desiccator
Style Handle type
Height (Metric) 386mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.