missing translation for 'onlineSavingsMsg'
Learn More

Acrylic acid, low water content, 99.5%, stab. with ca 200ppm 4-methoxyphenol, Thermo Scientific Chemicals

Product Code. 11470570
Click to view available options
Quantity:
500 mL
2.5 L

Acrylic acid acts as a precursor in the production of 3-hydroxypropionic acid. It is used in the preparation of water-absorbent resins. It reacts with alcohols to prepare the corresponding esters. Its esters are used as raw materials for synthetic resins, rubbers, coating adhesives, water-based paints, floor polishes and adhesives. It is also used to prepare forming homopolymers or copolymers by reacting with other monomers like acrylamides, acrylonitrile, vinyl, styrene, and butadiene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-10-7
Molecular Formula C3H4O2
Molecular Weight (g/mol) 72.063
MDL Number MFCD00004367
InChI Key NIXOWILDQLNWCW-UHFFFAOYSA-N
Synonym acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
PubChem CID 6581
ChEBI CHEBI:18308
IUPAC Name prop-2-enoic acid
SMILES C=CC(=O)O

Specifications

Melting Point ∼13°C
Density 1.051 g/mL
Boiling Point 138°C to 139°C
Flash Point 54°C (130°F)
Odor Pungent
Refractive Index 1.4202
Quantity 500 mL
UN Number UN2218
Beilstein 635743
Merck Index 14,130
Solubility Information Miscible with water.
Formula Weight 72.06
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material Acrylic acid, Low water content, Stabilized with ≈200ppm 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'